Learn More
N-(4-Bromobutyl)phthalimide, 96%
CAS: 5394-18-3 | C12H12BrNO2 | 282.14 g/mol
Supplier: Thermo Scientific Chemicals A1451714
Description
N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Store at room temperature. Store away from oxidizing agents.
Specifications
| N-(4-Bromobutyl)phthalimide | |
| 77°C to 81°C | |
| Odorless | |
| MFCD00005905 | |
| 164119 | |
| Soluble in ethanol. Insoluble in water. | |
| BrCCCCN1C(=O)C2=CC=CC=C2C1=O | |
| 282.14 | |
| 282.14 |
| 5394-18-3 | |
| 165°C to 170°C (1 mmHg) | |
| C12H12BrNO2 | |
| 25 g | |
| n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide | |
| UXFWTIGUWHJKDD-UHFFFAOYSA-N | |
| 2-(4-bromobutyl)isoindole-1,3-dione | |
| 93575 | |
| 98% |
Chemical Identifiers
| 5394-18-3 | |
| 282.14 | |
| UXFWTIGUWHJKDD-UHFFFAOYSA-N | |
| 93575 | |
| BrCCCCN1C(=O)C2=CC=CC=C2C1=O |
| C12H12BrNO2 | |
| MFCD00005905 | |
| n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide | |
| 2-(4-bromobutyl)isoindole-1,3-dione |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-401-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only