missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide 98.0+%, TCI America™

Catalog No. p-7137797
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35571G 1 g
T35575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35571G Supplier TCI America Supplier No. T35571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616880-14-9
Molecular Formula C13H20BNO4S
Molecular Weight (g/mol) 297.176
MDL Number MFCD05663876
InChI Key NPSPNWPZDXQYAC-UHFFFAOYSA-N
Synonym n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanesulfonamide, 4-methanesulfonylamino phenylboronic acid pinacol ester, 4-methanesulfonylaminophenylboronic acid, pinacol ester, 4-methanesulfonylaminophenylboronic acid pinacol ester, n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanesulfonamide, amtb416, 4-methanesulfonylamino phenyl boronic acid pinacol ester, 4-methanesulfonamido phenylboronic acid pinacol ester
PubChem CID 16217653
IUPAC Name N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NS(=O)(=O)C

Specifications

Melting Point 197°C
Color White
Quantity 1 g
Formula Weight 297.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide