missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(2-Benzimidazolyl)phenyl]maleimide 98.0+%, TCI America™

Catalog No. p-7118880
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B114950MG 50 mg
B1149100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B114950MG Supplier TCI America Supplier No. B114950MG
Only null left

Chemical Identifiers

CAS 27030-97-3
Molecular Formula C17H11N3O2
Molecular Weight (g/mol) 289.294
MDL Number MFCD00042767
InChI Key NZDOXVCRXDAVII-UHFFFAOYSA-N
PubChem CID 117909
IUPAC Name 1-[4-(1H-benzimidazol-2-yl)phenyl]pyrrole-2,5-dione
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)N4C(=O)C=CC4=O

Specifications

Melting Point 247°C
Color Yellow
Quantity 50 mg
Formula Weight 289.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[4-(2-Benzimidazolyl)phenyl]maleimide