accessibility menu, dialog, popup

UserName

CAS RN 27030-97-3

OONHNN

CAS RN 27030-97-3

IUPAC Name: 1-[4-(1H-1,3-benzodiazol-2-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[4-(1H-1,3-benzodiazol-2-yl)phenyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 289.29
Molecular Formula: C17H11N3O2
InChi Key: NZDOXVCRXDAVII-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=C(C=C1)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results