missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Pyridyl)pivalamide 98.0+%, TCI America™
Supplier: TCI America P210925G
Specifications
| N-(3-Pyridyl)pivalamide | |
| 130°C | |
| MFCD00996243 | |
| 2,2-dimethyl-n-pyridin-3-yl-propionamide, n-pyridin-3-yl pivalamide, 2,2-dimehtyl-n-pyridin-3-yl-propionamide, 2,2-dimethyl-n-pyridin-3-yl propanamide, 2,2-dimethyl-n-3-pyridyl propionamide, 2,2-dimethyl-n-pyridine-3yl-propionamide, 3-pivaloylamino pyridine, propanamide, 2,2-dimethyl-n-3-pyridinyl, 2,2-dimethyl-n-3-pyridinyl propanamide, n-3-pyridylpivalamide | |
| CC(C)(C)C(=O)NC1=CN=CC=C1 | |
| 178.235 | |
| 178.24 | |
| Crystalline Powder |
| 70298-88-3 | |
| C10H14N2O | |
| 25 g | |
| VQXVCVTZSTYIMG-UHFFFAOYSA-N | |
| 2,2-dimethyl-N-pyridin-3-ylpropanamide | |
| 4655044 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 70298-88-3 | |
| 178.235 | |
| VQXVCVTZSTYIMG-UHFFFAOYSA-N | |
| 4655044 | |
| CC(C)(C)C(=O)NC1=CN=CC=C1 |
| C10H14N2O | |
| MFCD00996243 | |
| 2,2-dimethyl-n-pyridin-3-yl-propionamide, n-pyridin-3-yl pivalamide, 2,2-dimehtyl-n-pyridin-3-yl-propionamide, 2,2-dimethyl-n-pyridin-3-yl propanamide, 2,2-dimethyl-n-3-pyridyl propionamide, 2,2-dimethyl-n-pyridine-3yl-propionamide, 3-pivaloylamino pyridine, propanamide, 2,2-dimethyl-n-3-pyridinyl, 2,2-dimethyl-n-3-pyridinyl propanamide, n-3-pyridylpivalamide | |
| 2,2-dimethyl-N-pyridin-3-ylpropanamide |
Safety and Handling
TSCA : No