accessibility menu, dialog, popup

UserName

CAS RN 70298-88-3

IUPAC Name: 2,2-dimethyl-N-(pyridin-3-yl)propanamide
Synonyms: 2,2-dimethyl-N-(pyridin-3-yl)propanamide
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: VQXVCVTZSTYIMG-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=CN=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results