missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Pyridyl)pivalamide 98.0+%, TCI America™

Catalog No. P21095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21095G 5 g
P210925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21095G Supplier TCI America Supplier No. P21095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70298-88-3
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00996243
InChI Key VQXVCVTZSTYIMG-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-3-yl-propionamide, n-pyridin-3-yl pivalamide, 2,2-dimehtyl-n-pyridin-3-yl-propionamide, 2,2-dimethyl-n-pyridin-3-yl propanamide, 2,2-dimethyl-n-3-pyridyl propionamide, 2,2-dimethyl-n-pyridine-3yl-propionamide, 3-pivaloylamino pyridine, propanamide, 2,2-dimethyl-n-3-pyridinyl, 2,2-dimethyl-n-3-pyridinyl propanamide, n-3-pyridylpivalamide
PubChem CID 4655044
IUPAC Name 2,2-dimethyl-N-pyridin-3-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CN=CC=C1

Specifications

Melting Point 130°C
Quantity 5 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Pyridyl)pivalamide