missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxypropyl)phthalimide, 98%

Catalog No. AAL0450709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0450709 10 g
AAL0450718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0450709 Supplier Thermo Scientific Chemicals Supplier No. L0450709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 883-44-3 | C11H11NO3 | 205.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 883-44-3
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00023097
InChI Key BSMILTTURCQDGJ-UHFFFAOYSA-N
Synonym n-3-hydroxypropyl phthalimide, 2-3-hydroxypropyl isoindoline-1,3-dione, n-3-hydroxypropyl-phthalimide, 1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl, 3-phthalimido-1-propanol, 2-3-hydroxypropyl isoindole-1,3-dione, 2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione, 3-phthalimidopropanol, 3-phthalimidylpropanol, acmc-209qsg
PubChem CID 70160
IUPAC Name 2-(3-hydroxypropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO

Specifications

Melting Point 75°C to 80°C
Quantity 10 g
Beilstein 158489
Formula Weight 205.22
Percent Purity 98%
Chemical Name or Material N-(3-Hydroxypropyl)phthalimide
EINECSNumber 212-931-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only