accessibility menu, dialog, popup

UserName

HONOO

CAS RN 883-44-3

IUPAC Name: 2-(3-hydroxypropyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(3-hydroxypropyl)isoindole-1,3-dione
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: BSMILTTURCQDGJ-UHFFFAOYSA-N
SMILES: OCCCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)