missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxyphenyl)thiourea, 97%

Catalog No. aal0997509
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0997509 10 g
AAL0997504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0997509 Supplier Thermo Scientific Chemicals Supplier No. L0997509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3394-05-6 | C8H8N2O6 | 228.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3394-05-6
MDL Number MFCD00022167

Specifications

Melting Point 175°C to 180°C
Odor Odorless
Molecular Formula C7H8N2OS
Quantity 10 g
Beilstein 2090372
Synonym 1-3-hydroxyphenyl-2-thiourea, 3-hydroxyphenyl thiourea, 1-3-hydroxyphenyl thiourea, 3-aminothioxomethyl amino phenol, acmc-20apbj, 1-m-hydroxyphenyl thiourea, 3-hydroxy-phenylthioharnstoff, thiourea, 3-hydroxyphenyl, thiourea,n-3-hydroxyphenyl, 1-m-hydroxyphenyl-2-thiourea
InChI Key BHJYKFUCQNISJA-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)O)NC(=S)N
IUPAC Name (3-hydroxyphenyl)thiourea
Molecular Weight (g/mol) 168.21
PubChem CID 2759330
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material N-(3-Hydroxyphenyl)thiourea
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only