missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxyphenyl)thiourea, 97%

Catalog No. AAL0997504
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0997504 2 g
AAL0997509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0997504 Supplier Thermo Scientific Chemicals Supplier No. L0997504
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3394-05-6 | C8H8N2O6 | 228.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3394-05-6
MDL Number MFCD00022167

Specifications

Melting Point 175°C to 180°C
Odor Odorless
Molecular Formula C8H8N2O6
Quantity 2 g
Beilstein 2090372
Synonym 4,5-dinitro veratrole, 1,2-dimethoxy-4,5-dinitro-benzene, 4,5-dimethoxy-1,2-dinitrobenzene, 4,5-dinitroveratrole, acmc-1ckho, ghl.pd_mitscher_leg0.933, benzene,1,2-dimethoxy-4,5-dinitro, benzene, 1,2-dimethoxy-4,5-dinitro
InChI Key WFDHPWTYKOAFBJ-UHFFFAOYSA-N
SMILES COC1=C(C=C(C(=C1)[N+](=O)[O-])[N+](=O)[O-])OC
IUPAC Name 1,2-dimethoxy-4,5-dinitrobenzene
Molecular Weight (g/mol) 228.16
PubChem CID 222989
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material N-(3-Hydroxyphenyl)thiourea
Show More Show Less
missing translation for 'healthHazard1' H302
missing translation for 'healthHazard2' GHS H Statement
H302
Harmful if swallowed.
missing translation for 'healthHazard3' P264b-P270-P301+P312-P330-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only