missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Chlorophenyl)-3-oxobutyramide 96.0+%, TCI America™
Supplier: TCI America C29841G
Specifications
| N-(3-Chlorophenyl)-3-oxobutyramide | |
| 103°C | |
| C10H10ClNO2 | |
| 1 g | |
| MTPKMGABYQNMMG-UHFFFAOYSA-N | |
| N-(3-chlorophenyl)-3-oxobutanamide | |
| 75487 | |
| ≥96.0% (HPLC,T) |
| 2415-87-4 | |
| White-Yellow | |
| MFCD00018440 | |
| 3′C-Chloroacetoacetanilide | |
| CC(=O)CC(=O)NC1=CC=CC(Cl)=C1 | |
| 211.65 | |
| 211.65 | |
| Crystalline Powder |
Chemical Identifiers
| 2415-87-4 | |
| 211.65 | |
| MTPKMGABYQNMMG-UHFFFAOYSA-N | |
| 75487 | |
| CC(=O)CC(=O)NC1=CC=CC(Cl)=C1 |
| C10H10ClNO2 | |
| MFCD00018440 | |
| 3′C-Chloroacetoacetanilide | |
| N-(3-chlorophenyl)-3-oxobutanamide |
Safety and Handling
EINECSNumber : (3)-0220
TSCA : Yes