missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Chlorophenyl)-3-oxobutyramide 96.0+%, TCI America™

Catalog No. p-7125763
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2984200MG 200 mg
C29841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2984200MG Supplier TCI America Supplier No. C2984200MG
Only null left

Chemical Identifiers

CAS 2415-87-4
Molecular Formula C10H10ClNO2
Molecular Weight (g/mol) 211.65
MDL Number MFCD00018440
InChI Key MTPKMGABYQNMMG-UHFFFAOYSA-N
Synonym 3′C-Chloroacetoacetanilide
PubChem CID 75487
IUPAC Name N-(3-chlorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC(Cl)=C1

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 200 mg
Formula Weight 211.65
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Chlorophenyl)-3-oxobutyramide
EINECSNumber (3)-0220
TSCA Yes