missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Chloro-2-methylphenyl)anthranilic Acid 97.0+%, TCI America™

Catalog No. C206425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C206425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C206425G Supplier TCI America Supplier No. C206425G

Chemical Identifiers

CAS 13710-19-5
Molecular Formula C14H12ClNO2
Molecular Weight (g/mol) 261.705
MDL Number MFCD00133865
InChI Key YEZNLOUZAIOMLT-UHFFFAOYSA-N
Synonym tolfenamic acid, clotam, n-3-chloro-2-methylphenyl anthranilic acid, 2-3-chloro-2-methylphenyl amino benzoic acid, tolfedine, acido tolfenamico, acide tolfenamique, acidum tolfenamicum, tolfine, n-2-methyl-3-chlorophenyl anthranilic acid
PubChem CID 610479
ChEBI CHEBI:32243
IUPAC Name 2-(3-chloro-2-methylanilino)benzoic acid
SMILES CC1=C(C=CC=C1Cl)NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 212°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 261.71
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Chloro-2-methylphenyl)anthranilic Acid
RTECSNumber CB2687500
TSCA No