missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Bromopropyl)phthalimide, 98%

Catalog No. AAA1558714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1558714 25 g
AAA1558722 100 g
AAA1558736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1558714 Supplier Thermo Scientific Chemicals Supplier No. A1558714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5460-29-7 | C11H10BrNO2 | 268.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5460-29-7
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD00005904
InChI Key VKJCJJYNVIYVQR-UHFFFAOYSA-N
Synonym n-3-bromopropyl phthalimide, 2-3-bromopropyl isoindoline-1,3-dione, 1-phthalimido-3-bromopropane, 3-bromopropylphthalimide, n-3-bromopropylphthalimide, gamma-bromopropylphthalimide, 2-3-bromopropyl-1h-isoindole-1,3 2h-dione, 2-3-bromopropyl-2,3-dihydro-1h-isoindole-1,3-dione, phthalimide, n-3-bromopropyl, 1h-isoindole-1,3 2h-dione, 2-3-bromopropyl
PubChem CID 21611
IUPAC Name 2-(3-bromopropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCBr

Specifications

Melting Point 72°C to 75°C
Odor Odorless
Quantity 25 g
Beilstein 157547
Formula Weight 268.12
Percent Purity 98%
Chemical Name or Material N-(3-Bromopropyl)phthalimide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-738-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only