missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Bromopropyl)phthalimide, 98%

Catalog No. p-76976
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107390250 25 g Glass bottle
AC107391000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107390250 Supplier Thermo Scientific Chemicals Supplier No. 107390250
Only null left

CAS: 5460-29-7 | C11H10BrNO2 | 268.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5460-29-7
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.1
MDL Number MFCD00005904
InChI Key VKJCJJYNVIYVQR-UHFFFAOYSA-N
Synonym n-3-bromopropyl phthalimide, 2-3-bromopropyl isoindoline-1,3-dione, 1-phthalimido-3-bromopropane, 3-bromopropylphthalimide, n-3-bromopropylphthalimide, gamma-bromopropylphthalimide, 2-3-bromopropyl-1h-isoindole-1,3 2h-dione, 2-3-bromopropyl-2,3-dihydro-1h-isoindole-1,3-dione, phthalimide, n-3-bromopropyl, 1h-isoindole-1,3 2h-dione, 2-3-bromopropyl
PubChem CID 21611
IUPAC Name 2-(3-bromopropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 76°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 462
Solubility Information Solubility in water: <1g/L. Other solubilities: soluble ca. 10g/L in organic solvents,soluble in ethanol and ether
Formula Weight 268.1
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-(3-Bromopropyl)phthalimide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 226-738-5

RUO – Research Use Only