missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)-2-nitrobenzenesulfonamide Hydrochloride 98.0+%, TCI America™

Catalog No. A22685G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22685G 5 g
A22681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22685G Supplier TCI America Supplier No. A22685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 863983-46-4
Molecular Formula C9H14ClN3O4S
Molecular Weight (g/mol) 295.74
MDL Number MFCD07366920
InChI Key UADJAJMVAIAKHQ-UHFFFAOYSA-N
Synonym 1-Amino-3-(2-nitrobenzenesulfonamido)propane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,3-propanediamine Hydrochloride, N-(o-Ns)-1,3-diaminopropane Hydrochloride
PubChem CID 44630012
IUPAC Name N-(3-aminopropyl)-2-nitrobenzene-1-sulfonamide hydrochloride
SMILES Cl.NCCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 295.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Aminopropyl)-2-nitrobenzenesulfonamide Hydrochloride