missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine, 97%

 

Supplier:  Thermo Scientific™ 333915000

Catalog No. AC333915000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine
58970-76-6
100.0
96% min. (HPLC)
C16H24N2O4
500mg
bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
VGGGPCQERPFHOB-MCIONIFRSA-N
(2S)-2-[[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl]amino]-4-methylpentanoic acid
72172
97%
97%
96.0
Authentic
Glass bottle
MFCD00083262
15, 10024
Solubility in water: slightly soluble. Other solubilities: soluble methanol, ethanol,, soluble in acetic acid, aq. hcl, alkaline solution
CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O
308.38
308.38

Chemical Identifiers

58970-76-6
308.38
VGGGPCQERPFHOB-MCIONIFRSA-N
72172
CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O
C16H24N2O4
MFCD00083262
bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
(2S)-2-[[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl]amino]-4-methylpentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 261-529-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only