missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ N-2 Supplement (100X)

Catalog No. 17502001
Encompass
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17502001 50 mL
17502048 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17502001 Supplier Gibco™ Supplier No. 17502001
Only null left

N-2 Supplement is a versatile serum-free cell culture supplement that supports the growth and viability of various cell types.

Gibco N-2 Supplement is a chemically defined, serum-free supplement based on Bottenstein's N-1 formulation, containing optimized concentrations of insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite. N-2 Supplement is recommended for growth and expression of neuroblastomas and neural cells in primary cultures from both the peripheral nervous system (PNS) and the central nervous system (CNS).

N-2 Supplement has been demonstrated in wide-ranging applications, such as the culture of neural stem cells, the culture of the SH-SY5Y cell line (subline of the neuroblastoma cell line SK-N-SH), and culture of the HT22 mouse hippocampal neuronal cell line. Additionally, N-2 Supplement adds support to cell culture media used to differentiate pluripotent stem cells (PSCs) to a variety of cell types, including neural and pancreatic-like cell types.

N-2 Supplement may be used with Neurobasal media supplemented with growth factors such as bFGF and EGF. When combined with Dulbecco's Modified Eagle Medium (D-MEM) and fetal bovine serum (FBS), N-2 Supplement also provides suitable supplementation for the culture of human and rodent astrocytes.

  • Each lot is cGMP manufactured
  • Each lot is tested for sterility, endotoxin, and performance
  • Contains insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite
  • Can be used as a substitute for Bottenstein's N-1 formulation
  • Provided as a 100X concentrate

Choose Gibco N-2 Supplement for a reliable, high-quality product trusted by researchers worldwide.

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.

Cell Type Neural Cells
Classification Serum-free
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Quantity 50 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.