missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Pyridylmethyl)glycine Ethyl Ester 95.0+%, TCI America™

Catalog No. p-7134999
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
P11941G 1 g
P11945G 5 g
2 options

Catalog No. P11941G

Supplier: TCI America P11941G

Only null left

Chemical Identifiers

CAS 62402-24-8
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD00191573
InChI Key UCSBXEQEHOOKHG-UHFFFAOYSA-N
Synonym Ethyl N-(2-Pyridylmethyl)glycinate, N-(2-Picolyl)glycine Ethyl Ester
PubChem CID 13236080
IUPAC Name ethyl 2-(pyridin-2-ylmethylamino)acetate
SMILES CCOC(=O)CNCC1=CC=CC=N1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 194.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Pyridylmethyl)glycine Ethyl Ester