accessibility menu, dialog, popup

UserName

H3COONHN

CAS RN 62402-24-8

IUPAC Name: ethyl 2-{[(pyridin-2-yl)methyl]amino}acetate
Synonyms: ethyl 2-[(pyridin-2-ylmethyl)amino]acetate
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: UCSBXEQEHOOKHG-UHFFFAOYSA-N
SMILES: CCOC(=O)CNCC1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results