missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Pyridyl)thiourea, 98%

Catalog No. p-7045645
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1146503 1 g
AAL1146506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1146503 Supplier Thermo Scientific Chemicals Supplier No. L1146503
Only null left

CAS: 14294-11-2 | C6H7N3S | 153.20 g/mol

N-(2-Pyridyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Pyridyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 14294-11-2
Molecular Formula C6H7N3S
Molecular Weight (g/mol) 153.20
MDL Number MFCD00041227
InChI Key SLUHLANJIVXTRQ-UHFFFAOYSA-N
Synonym 1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea
PubChem CID 1490491
IUPAC Name pyridin-2-ylthiourea
SMILES NC(=S)NC1=CC=CC=N1

Specifications

Melting Point 146°C to 149°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 117849
Formula Weight 153.21
Percent Purity 98%
Chemical Name or Material N-(2-Pyridyl)thiourea
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only