missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Pyridyl)pivalamide 98.0+%, TCI America™

Catalog No. p-7135554
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23855G 5 g
P238525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23855G Supplier TCI America Supplier No. P23855G

Chemical Identifiers

CAS 86847-59-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00460277
InChI Key CGSPVYCZBDFPHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-2-yl-propionamide, 2-pivaloylamino pyridine, 2,2-dimethyl-n-pyridin-2-yl propanamide, n-pyridin-2-yl pivalamide, n-boc-2-aminopyridine, 2,2-dimethyl-n-2-pyridyl propanamide, 2,2-dimethyl-n-2-pyridinyl propanamide, propanamide, 2,2-dimethyl-n-2-pyridinyl, pivaloylamino pyridine, n-o-pyridylpivalamide
PubChem CID 1051519
IUPAC Name 2,2-dimethyl-N-pyridin-2-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=CC=N1

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Pyridyl)pivalamide