missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Phenylacetyl)glycine, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014341
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014341 250 mg
111014339 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014341 Supplier Merck Emd Millipore Supplier No. 96408250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 500-98-1
Linear Formula C10H11 NO3
MDL Number MFCD00021744
Synonym 2-(2-Phenylacetamido)acetic acid; 2-Phenylacetyl glycine; NSC 408424; NSC 92778; Phenaceturic acid; Phenacetylglycine
Type Analytical Standard
Quantity 250 mg
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard