missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxyethyl)propylamine 98.0+%, TCI America™

Catalog No. M09805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09805G 5 g
M098025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09805G Supplier TCI America Supplier No. M09805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43175-57-1
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00144830
InChI Key UDZCEFCJEGGQOJ-UHFFFAOYSA-N
PubChem CID 12238049
IUPAC Name N-(2-methoxyethyl)propan-1-amine
SMILES CCCNCCOC

Specifications

Color Colorless
Boiling Point 138°C
Quantity 5 g
UN Number 2733
Formula Weight 117.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Methoxyethyl)propylamine