accessibility menu, dialog, popup

UserName

H3CHNOCH3

CAS RN 43175-57-1

IUPAC Name: (2-methoxyethyl)(propyl)amine
Synonyms: (2-methoxyethyl)(propyl)amine
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: UDZCEFCJEGGQOJ-UHFFFAOYSA-N
SMILES: CCCNCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results