missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxyethyl)methylamine 97.0+%, TCI America™

Catalog No. p-7132478
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09865ML 5 mL
M098625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09865ML Supplier TCI America Supplier No. M09865ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38256-93-8
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00144829
InChI Key KOHBEDRJXKOYHL-UHFFFAOYSA-N
PubChem CID 300977
IUPAC Name 2-methoxy-N-methylethanamine
SMILES CNCCOC

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 mL
UN Number 2924
Formula Weight 89.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Methoxyethyl)methylamine