accessibility menu, dialog, popup

UserName

CAS RN 38256-93-8

IUPAC Name: (2-methoxyethyl)(methyl)amine
Synonyms: (2-methoxyethyl)(methyl)amine
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: KOHBEDRJXKOYHL-UHFFFAOYSA-N
SMILES: CNCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results