missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxyethyl)ethylamine 97.0+%, TCI America™

Catalog No. E03975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03975G 5 g
E039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03975G Supplier TCI America Supplier No. E03975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34322-82-2
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00144876
InChI Key VGEMYWDUTPQWBN-UHFFFAOYSA-N
Synonym N-Ethyl-2-methoxyethylamine
PubChem CID 546877
IUPAC Name ethyl(2-methoxyethyl)amine
SMILES CCNCCOC

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
UN Number 2734
Formula Weight 103.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Methoxyethyl)ethylamine