accessibility menu, dialog, popup

UserName

H3CHNOCH3

CAS RN 34322-82-2

IUPAC Name: ethyl(2-methoxyethyl)amine
Synonyms: ethyl(2-methoxyethyl)amine
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: VGEMYWDUTPQWBN-UHFFFAOYSA-N
SMILES: CCNCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results