missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Hydroxyethyl)phthalimide, 98+%
CAS: 3891-07-4 | C17H22NO6 | 336.37 g/mol
Supplier: Thermo Scientific Chemicals A1425936
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-(2-Hydroxyethyl)phthalimide | |
| 126°C to 129°C | |
| MFCD00005903 | |
| 147214 | |
| GLMODRZPPBZPPB-ZDUSSCGKSA-M | |
| (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid | |
| 107472 | |
| ≥98% |
| 3891-07-4 | |
| C17H22NO6 | |
| 500 g | |
| z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261 | |
| CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 336.37 | |
| 191.19 |
Chemical Identifiers
| 3891-07-4 | |
| MFCD00005903 |
Safety and Handling
EINECSNumber : 223-434-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only