missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide, 98+%

Catalog No. p-7021783
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1425922 100 g
AAA1425936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1425922 Supplier Thermo Scientific Chemicals Supplier No. A1425922
Only null left

CAS: 3891-07-4 | C17H22NO6 | 336.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3891-07-4
MDL Number MFCD00005903

Specifications

Melting Point 126°C to 129°C
Molecular Formula C14H19NO3
Quantity 100 g
Beilstein 147214
Synonym 2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, n-pinacolylphthalamic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, +-n-pinacolylphthalamic acid
InChI Key QUZGNIZLRAGRHB-UHFFFAOYNA-N
SMILES CC(C(C)(C)C)NC(=O)C1=CC=CC=C1C(=O)O
IUPAC Name 2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
Molecular Weight (g/mol) 249.31
PubChem CID 3354762
Formula Weight 191.19
Percent Purity ≥98%
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less
EINECSNumber 223-434-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only