missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Hydroxyethyl)iminodiacetic Acid 98.0+%, TCI America™
$83.89 - $279.60
Chemical Identifiers
| CAS | 93-62-9 |
|---|---|
| Molecular Formula | C6H11NO5 |
| Molecular Weight (g/mol) | 177.156 |
| MDL Number | MFCD00004293 |
| InChI Key | JYXGIOKAKDAARW-UHFFFAOYSA-N |
| Synonym | n-2-hydroxyethyl iminodiacetic acid, heida, ethanolamine-n,n-diacetic acid, hydroxyethylimino diacetic acid, 2-hydroxyethylaminodiacetic acid, glycine, n-carboxymethyl-n-2-hydroxyethyl, usaf do-37, acetic acid, hydroxyethyl iminodi, hydroxyethylnitrilodiacetic acid, n-hydroxyethyliminodiacetic acid |
| PubChem CID | 7152 |
| IUPAC Name | 2-[carboxymethyl(2-hydroxyethyl)amino]acetic acid |
| SMILES | C(CO)N(CC(=O)O)CC(=O)O |
Chemical Identifiers
| 93-62-9 | |
| 177.156 | |
| JYXGIOKAKDAARW-UHFFFAOYSA-N | |
| 7152 | |
| C(CO)N(CC(=O)O)CC(=O)O |
| C6H11NO5 | |
| MFCD00004293 | |
| n-2-hydroxyethyl iminodiacetic acid, heida, ethanolamine-n,n-diacetic acid, hydroxyethylimino diacetic acid, 2-hydroxyethylaminodiacetic acid, glycine, n-carboxymethyl-n-2-hydroxyethyl, usaf do-37, acetic acid, hydroxyethyl iminodi, hydroxyethylnitrilodiacetic acid, n-hydroxyethyliminodiacetic acid | |
| 2-[carboxymethyl(2-hydroxyethyl)amino]acetic acid |
Specifications
| 93-62-9 | |
| White | |
| MFCD00004293 | |
| n-2-hydroxyethyl iminodiacetic acid, heida, ethanolamine-n,n-diacetic acid, hydroxyethylimino diacetic acid, 2-hydroxyethylaminodiacetic acid, glycine, n-carboxymethyl-n-2-hydroxyethyl, usaf do-37, acetic acid, hydroxyethyl iminodi, hydroxyethylnitrilodiacetic acid, n-hydroxyethyliminodiacetic acid | |
| C(CO)N(CC(=O)O)CC(=O)O | |
| 177.156 | |
| 177.16 | |
| Crystalline Powder |
| 177°C | |
| C6H11NO5 | |
| 5 g | |
| JYXGIOKAKDAARW-UHFFFAOYSA-N | |
| 2-[carboxymethyl(2-hydroxyethyl)amino]acetic acid | |
| 7152 | |
| ≥98.0% (T) | |
| N-(2-Hydroxyethyl)iminodiacetic Acid |
Safety and Handling
EINECSNumber : (2)-1282
RTECSNumber : AI1925000
TSCA : Yes