missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)iminodiacetic Acid 98.0+%, TCI America™

Catalog No. H048925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H048925G 25 g
H04895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H048925G Supplier TCI America Supplier No. H048925G
Only null left

Chemical Identifiers

CAS 93-62-9
Molecular Formula C6H11NO5
Molecular Weight (g/mol) 177.156
MDL Number MFCD00004293
InChI Key JYXGIOKAKDAARW-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl iminodiacetic acid, heida, ethanolamine-n,n-diacetic acid, hydroxyethylimino diacetic acid, 2-hydroxyethylaminodiacetic acid, glycine, n-carboxymethyl-n-2-hydroxyethyl, usaf do-37, acetic acid, hydroxyethyl iminodi, hydroxyethylnitrilodiacetic acid, n-hydroxyethyliminodiacetic acid
PubChem CID 7152
IUPAC Name 2-[carboxymethyl(2-hydroxyethyl)amino]acetic acid
SMILES C(CO)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 177°C
Color White
Quantity 25 g
Formula Weight 177.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)iminodiacetic Acid
EINECSNumber (2)-1282
RTECSNumber AI1925000
TSCA Yes