missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)formamide, 97%

CAS: 693-06-1 | C3H7NO2 | 89.09 g/mol

Supplier:  Thermo Scientific Chemicals H5234503

 View more versions of this product

Catalog No. AAH5234503

Only null left
Explore available promotions

Description

Description

N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

N-(2-Hydroxyethyl)formamide
C3H7NO2
MFCD00021040
n-2-hydroxyethyl formamide, formamide, n-2-hydroxyethyl, n-formylethanolamine
OCCNC=O
89.09
89.09
693-06-1
HCONHCH2CH2OH
1 g
BAMUPQJDKBGDPU-UHFFFAOYSA-N
N-(2-hydroxyethyl)formamide
69657
97%

Chemical Identifiers

693-06-1
89.09
BAMUPQJDKBGDPU-UHFFFAOYSA-N
69657
OCCNC=O
C3H7NO2
MFCD00021040
n-2-hydroxyethyl formamide, formamide, n-2-hydroxyethyl, n-formylethanolamine
N-(2-hydroxyethyl)formamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-741-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only