missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)formamide, 97%

Catalog No. p-7034653
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52345MD 250 mg
AAH5234503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52345MD Supplier Thermo Scientific Chemicals Supplier No. H52345MD
Only null left

CAS: 693-06-1 | C3H7NO2 | 89.09 g/mol

N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 693-06-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00021040
InChI Key BAMUPQJDKBGDPU-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl formamide, formamide, n-2-hydroxyethyl, n-formylethanolamine
PubChem CID 69657
IUPAC Name N-(2-hydroxyethyl)formamide
SMILES OCCNC=O

Specifications

Linear Formula HCONHCH2CH2OH
Quantity 250 mg
Formula Weight 89.09
Percent Purity 97%
Chemical Name or Material N-(2-Hydroxyethyl)formamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-741-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only