missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic Acid 98.0+%, TCI America™

Catalog No. h0243100g
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0243100G 100 g
H024325G 25 g
H0243500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0243100G Supplier TCI America Supplier No. H0243100G
Only null left

Chemical Identifiers

CAS 150-39-0
Molecular Formula C10H15N2Na3O7
Molecular Weight (g/mol) 344.21
MDL Number MFCD00004294
InChI Key WHNXAQZPEBNFBC-UHFFFAOYSA-K
Synonym hedta, heedta, versenol, detarol, n-2-hydroxyethyl ethylenediaminetriacetic acid, chel dm acid, versenol 120, hamp-ol acid, 2-hydroxyethyl ethylenediaminetriacetic acid, oxyethylethylenediaminetriacetic acid
PubChem CID 8773
IUPAC Name trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(2-hydroxyethyl)amino)acetate
SMILES [Na+].[Na+].[Na+].OCCN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 100 g
Formula Weight 278.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic Acid
RTECSNumber MB9185000
TSCA Yes