missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[2-(Dimethylamino)ethyl]-N-methylformamide 97.0+%, TCI America™
$130.92 - $390.80
Chemical Identifiers
| CAS | 105669-53-2 |
|---|---|
| Molecular Formula | C6H14N2O |
| Molecular Weight (g/mol) | 130.191 |
| MDL Number | MFCD04039895 |
| InChI Key | CSLBJRKWKVBRSQ-UHFFFAOYSA-N |
| PubChem CID | 279482 |
| IUPAC Name | N-[2-(dimethylamino)ethyl]-N-methylformamide |
| SMILES | CN(C)CCN(C)C=O |
Chemical Identifiers
| 105669-53-2 | |
| 130.191 | |
| CSLBJRKWKVBRSQ-UHFFFAOYSA-N | |
| N-[2-(dimethylamino)ethyl]-N-methylformamide |
| C6H14N2O | |
| MFCD04039895 | |
| 279482 | |
| CN(C)CCN(C)C=O |
Specifications
| 105669-53-2 | |
| 96°C | |
| MFCD04039895 | |
| N-Formyl-N,N′C,N′C-trimethylethylenediamine | |
| CN(C)CCN(C)C=O | |
| 130.191 | |
| 130.19 | |
| Liquid |
| Yellow | |
| C6H14N2O | |
| 5 g | |
| CSLBJRKWKVBRSQ-UHFFFAOYSA-N | |
| N-[2-(dimethylamino)ethyl]-N-methylformamide | |
| 279482 | |
| ≥97.0% (GC,T) | |
| N-[2-(Dimethylamino)ethyl]-N-methylformamide |
Safety and Handling
TSCA : No