missing translation for 'onlineSavingsMsg'
Learn More

N-[2-(Dimethylamino)ethyl]-N-methylformamide 97.0+%, TCI America™

Catalog No. d37245g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37245G 5 g
D372425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37245G Supplier TCI America Supplier No. D37245G
Only null left

Chemical Identifiers

CAS 105669-53-2
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD04039895
InChI Key CSLBJRKWKVBRSQ-UHFFFAOYSA-N
PubChem CID 279482
IUPAC Name N-[2-(dimethylamino)ethyl]-N-methylformamide
SMILES CN(C)CCN(C)C=O

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Synonym N-Formyl-N,N′C,N′C-trimethylethylenediamine
Formula Weight 130.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-[2-(Dimethylamino)ethyl]-N-methylformamide