missing translation for 'onlineSavingsMsg'
Learn More

N,2-Dimethyl-(1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC05746DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC05746DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

144163-81-5
142.22
GJHOBSIOQKGKSG-UHFFFAOYSA-N
18939489
CC1=NC(=CS1)CNC
C6H10N2S
MFCD06738964
methyl-2-methyl-thiazol-4-ylmethyl-amine, methyl-2-methylthiazole-4-yl methyl amine, n-methyl-1-2-methylthiazol-4-yl methanamine, methyl 2-methyl-1,3-thiazol-4-yl methyl amine, 4-thiazolemethanamine,n,2-dimethyl, n-methyl 2-methylthiazol-4-yl methanamine, 4-thiazolemethanamine, n,2-dimethyl, n-methyl-1-2-methyl-1,3-thiazol-4-yl methanamine, n,2-dimethyl-1,3-thiazol-4-yl methylamine, 2-methyl-4-methyl aminomethyl-1,3-thiazole
N-methyl-1-(2-methyl-1,3-thiazol-4-yl)methanamine

Specifications

N, 2-dimethyl-(1, 3-thiazol-4-yl)methylamine
96.5
97%
C6H10N2S
MFCD06738964
GJHOBSIOQKGKSG-UHFFFAOYSA-N
N-methyl-1-(2-methyl-1,3-thiazol-4-yl)methanamine
18939489
97%
144163-81-5
100.0
Amber Glass Bottle
1g
methyl-2-methyl-thiazol-4-ylmethyl-amine, methyl-2-methylthiazole-4-yl methyl amine, n-methyl-1-2-methylthiazol-4-yl methanamine, methyl 2-methyl-1,3-thiazol-4-yl methyl amine, 4-thiazolemethanamine,n,2-dimethyl, n-methyl 2-methylthiazol-4-yl methanamine, 4-thiazolemethanamine, n,2-dimethyl, n-methyl-1-2-methyl-1,3-thiazol-4-yl methanamine, n,2-dimethyl-1,3-thiazol-4-yl methylamine, 2-methyl-4-methyl aminomethyl-1,3-thiazole
CC1=NC(=CS1)CNC
142.22
142.23
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions