accessibility menu, dialog, popup

UserName

CAS RN 144163-81-5

CH3NSHNCH3

CAS RN 144163-81-5

IUPAC Name: methyl[(2-methyl-1,3-thiazol-4-yl)methyl]amine
Synonyms: methyl[(2-methyl-1,3-thiazol-4-yl)methyl]amine
Molecular Weight (g/mol): 142.22
Molecular Formula: C6H10N2S
InChi Key: GJHOBSIOQKGKSG-UHFFFAOYSA-N
SMILES: CNCC1=CSC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)