missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)-N-ethylaniline 98.0+%, TCI America™

Catalog No. p-7124430
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C045125G 25 g
C0451500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C045125G Supplier TCI America Supplier No. C045125G

Chemical Identifiers

CAS 148-87-8
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD00019858
InChI Key WYRNRZQRKCXPLA-UHFFFAOYSA-N
Synonym 3-(N-Ethyl-N-phenylamino)propionitrile
PubChem CID 8995
IUPAC Name 3-(N-ethylanilino)propanenitrile
SMILES CCN(CCC#N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 176°C
Quantity 25 g
Formula Weight 174.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Cyanoethyl)-N-ethylaniline
EINECSNumber (3)-0162
RTECSNumber BY0100000
TSCA Yes