accessibility menu, dialog, popup

UserName

CAS RN 148-87-8

H3CNN

CAS RN 148-87-8

IUPAC Name: 3-[ethyl(phenyl)amino]propanenitrile
Synonyms: 3-[ethyl(phenyl)amino]propanenitrile
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: WYRNRZQRKCXPLA-UHFFFAOYSA-N
SMILES: CCN(CCC#N)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results