missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Cyanoethyl)-N-ethyl-m-toluidine 98.0+%, TCI America™
Supplier: TCI America C28775G
Specifications
| N-(2-Cyanoethyl)-N-ethyl-m-toluidine | |
| Yellow | |
| MFCD00019859 | |
| 3-[Ethyl(3-methylphenyl)amino]propionitrile, 3-(N-Ethyl-m-toluidino)propionitrile | |
| CCN(CCC#N)C1=CC=CC(=C1)C | |
| 188.274 | |
| 188.27 | |
| Liquid |
| 148-69-6 | |
| C12H16N2 | |
| 5 g | |
| NPCCPMHHNIOSHL-UHFFFAOYSA-N | |
| 3-(N-ethyl-3-methylanilino)propanenitrile | |
| 67363 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 148-69-6 | |
| 188.274 | |
| NPCCPMHHNIOSHL-UHFFFAOYSA-N | |
| 67363 | |
| CCN(CCC#N)C1=CC=CC(=C1)C |
| C12H16N2 | |
| MFCD00019859 | |
| 3-[Ethyl(3-methylphenyl)amino]propionitrile, 3-(N-Ethyl-m-toluidino)propionitrile | |
| 3-(N-ethyl-3-methylanilino)propanenitrile |
Safety and Handling
EINECSNumber : (3)-0202
RTECSNumber : TZ4695000
TSCA : Yes
Recommended Storage : Refrigerator