missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)-N-ethyl-m-toluidine 98.0+%, TCI America™

Catalog No. c28771g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28771G 1 g
C28775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28771G Supplier TCI America Supplier No. C28771G

Chemical Identifiers

CAS 148-69-6
Molecular Formula C12H16N2
Molecular Weight (g/mol) 188.274
MDL Number MFCD00019859
InChI Key NPCCPMHHNIOSHL-UHFFFAOYSA-N
Synonym 3-[Ethyl(3-methylphenyl)amino]propionitrile, 3-(N-Ethyl-m-toluidino)propionitrile
PubChem CID 67363
IUPAC Name 3-(N-ethyl-3-methylanilino)propanenitrile
SMILES CCN(CCC#N)C1=CC=CC(=C1)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 188.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Cyanoethyl)-N-ethyl-m-toluidine
EINECSNumber (3)-0202
RTECSNumber TZ4695000
TSCA Yes
Recommended Storage Refrigerator