missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline 96.0+%, TCI America™

Catalog No. C045225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C045225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C045225G Supplier TCI America Supplier No. C045225G
Only null left

Chemical Identifiers

CAS 92-64-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD00035705
InChI Key WZJJWQVBLSPALW-UHFFFAOYSA-N
Synonym n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino
PubChem CID 113349
IUPAC Name 3-[N-(2-hydroxyethyl)anilino]propanenitrile
SMILES C1=CC=C(C=C1)N(CCC#N)CCO

Specifications

Color Yellow
Boiling Point 187°C
Quantity 25 g
Formula Weight 190.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline
EINECSNumber (3)-0161
RTECSNumber UG2905000
TSCA Yes