accessibility menu, dialog, popup

UserName

CAS RN 92-64-8

CH3ONHN

CAS RN 92-64-8

IUPAC Name: 3-[(2-ethoxyphenyl)amino]propanenitrile
Synonyms: 3-[(2-ethoxyphenyl)amino]propanenitrile
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: VATFTWNXRDYZLE-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1NCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results