missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Chlorophenyl)maleimide, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 1203-24-3 |
|---|---|
| Molecular Formula | C10H6ClNO2 |
| Molecular Weight (g/mol) | 207.62 |
| InChI Key | KPQOXMCRYWDRSB-UHFFFAOYSA-N |
| Synonym | 1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide |
| PubChem CID | 14550 |
| IUPAC Name | 1-(2-chlorophenyl)pyrrole-2,5-dione |
| SMILES | C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC457330010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 1203-24-3 | |
| 207.62 | |
| 1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide | |
| 1-(2-chlorophenyl)pyrrole-2,5-dione |
| C10H6ClNO2 | |
| KPQOXMCRYWDRSB-UHFFFAOYSA-N | |
| 14550 | |
| C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl |
Specifications
| 1203-24-3 | |
| 96% min. (GC) | |
| C10H6ClNO2 | |
| KPQOXMCRYWDRSB-UHFFFAOYSA-N | |
| 1-(2-chlorophenyl)pyrrole-2,5-dione | |
| 14550 | |
| 97% |
| Authentic | |
| Glass Bottle | |
| 1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide | |
| C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl | |
| 207.62 | |
| 207.62 | |
| N-(2-Chlorophenyl)maleimide |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 214-869-
RUO – Research Use Only