Learn More
N-(2-Chlorophenyl)maleimide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 457330010
| Quantity | 1g |
|---|
Chemical Identifiers
| 1203-24-3 | |
| 207.62 | |
| 1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide | |
| 1-(2-chlorophenyl)pyrrole-2,5-dione |
| C10H6ClNO2 | |
| KPQOXMCRYWDRSB-UHFFFAOYSA-N | |
| 14550 | |
| C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl |
Specifications
| N-(2-Chlorophenyl)maleimide | |
| Authentic | |
| Glass Bottle | |
| 1g | |
| KPQOXMCRYWDRSB-UHFFFAOYSA-N | |
| 1-(2-chlorophenyl)pyrrole-2,5-dione | |
| 14550 | |
| 97% |
| 1203-24-3 | |
| 96% min. (GC) | |
| C10H6ClNO2 | |
| 1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide | |
| C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl | |
| 207.62 | |
| 207.62 |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 214-869-