missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)morpholine hydrochloride, 99%

Catalog No. AC109200050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109200050 5 g
AC109201000 100 g
AC109205000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC109200050 Supplier Thermo Scientific Chemicals Supplier No. 109200050

CAS: 3647-69-6 | C6H12ClNO·HCl | 186.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3647-69-6
Molecular Formula C6H12ClNO·HCl
Molecular Weight (g/mol) 186.08
MDL Number MFCD00012797
InChI Key NBJHDLKSWUDGJG-UHFFFAOYSA-N
Synonym 4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride
PubChem CID 77210
IUPAC Name 4-(2-chloroethyl)morpholine;hydrochloride
SMILES C1COCCN1CCCl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 182°C to 185°C
Color White to Beige
Loss on Drying 1% max. (80°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 186.08
Percent Purity 99%
Physical Form Hygroscopic Crystalline Powder
Chemical Name or Material N-(2-Chloroethyl)morpholine hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-881-2
RTECSNumber QE0260000
TSCA TSCA

RUO – Research Use Only